MilliporeSigma
  • HmuY is an important virulence factor for Porphyromonas gingivalis growth in the heme-limited host environment and infection of macrophages.
HmuY is an important virulence factor for Porphyromonas gingivalis growth in the heme-limited host environment and infection of macrophages.
Biochemical and biophysical research communications (2015-10-21)
Teresa Olczak, Paulina Sosicka, Mariusz Olczak
ABSTRACT
Porphyromonas gingivalis, the main etiologic agent and key pathogen responsible for initiation and progression of chronic periodontitis, is a haem auxotroph, and the uptake of this compound is essential for its survival and the ability to establish an infection. The aim of this study was to examine the role of a hemophore-like HmuY protein in P. gingivalis growth and infection of macrophages. Inactivation of the hmuY gene caused reduced P. gingivalis growth in vitro in the presence of serum as a heme sole source, as well as in vivo co-cultures with THP-1-derived macrophages. This resulted in diminished invasion efficiency of macrophages by live bacteria lacking functional hmuY gene. Both features were partially restored after addition of the purified HmuY protein, which was internalized when added either together with the hmuY mutant strain or alone to macrophage cultures. We conclude that HmuY is an important virulence factor of P. gingivalis for infection of macrophages in a heme-limited host environment.
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Glutamine
SAFC
L-Cysteine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source