MilliporeSigma
  • Effects of Solute-Solvent Hydrogen Bonding on Nonaqueous Electrolyte Structure.
Effects of Solute-Solvent Hydrogen Bonding on Nonaqueous Electrolyte Structure.
The journal of physical chemistry letters (2015-08-13)
Kjell W Schroder, Anthony G Dylla, Logan D C Bishop, Elizabeth R Kamilar, Jennette Saunders, Lauren J Webb, Keith J Stevenson
ABSTRACT
We investigate the source of Raman background signal commonly misidentified as fluorescence in nonaqueous electrolytes via a variety of spectroscopies (Raman, fluorescence, NMR) and find evidence of hydrogen-bonding interactions. This hydrogen bonding gives rise to broadband anharmonic vibrational modes and suggests that anions play an important and underappreciated role in the structure of nonaqueous electrolytes. Controlling electrolyte structure has important applications in advancing in operando spectroscopy measurements as well as understanding the stability of high concentration electrolytes for next-generation electrochemical energy storage devices.
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Ethylene carbonate, 98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Diethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Lithium bis(oxalato)borate
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Diethyl carbonate, 99%