MilliporeSigma
  • MicroRNA-205 inhibits the proliferation and invasion of breast cancer by regulating AMOT expression.
MicroRNA-205 inhibits the proliferation and invasion of breast cancer by regulating AMOT expression.
Oncology reports (2015-08-05)
Haige Zhang, Qingxia Fan
ABSTRACT
It has been reported that the expression of angiomotin (AMOT) is upregulated in breast cancer. However, the regulatory mechanism remains unknown. In the present study, we aimed to ascertain whether the expression of AMOT is regulated by microRNAs (miRNAs) in breast cancer. In the present study, miR-205 was significantly downregulated in breast cancer samples and it was identified to directly target the 3'-untranslated region (3'-UTR) of AMOT in breast cancer MCF-7 cells by luciferase assay. miR-205 and small interfering RNA (siRNA)-mediated AMOT-knockdown experiments revealed that miR-205 significantly inhibited the proliferation and the invasion of MCF-7 cells through a decrease in the expression of AMOT, yet had no effect on apoptosis. Furthermore, we observed that the overexpression of AMOT partially reversed the inhibitory effect of miR-205 on the growth and the invasion of MCF-7 cells. The data indicated that miR-205 regulated the proliferation and the invasion of breast cancer cells through suppression of AMOT expression, at least partly. Therefore, the disordered decreased expression of miR-205 and the resulting AMOT upregulation contributes to breast carcinogenesis, and miR-205-AMOT represents a new potential therapeutic target for the treatment of breast carcinoma.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Amot
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human AMOT
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%