MilliporeSigma
  • Cytotoxicity of the Urokinase-Plasminogen Activator Inhibitor Carbamimidothioic Acid (4-Boronophenyl) Methyl Ester Hydrobromide (BC-11) on Triple-Negative MDA-MB231 Breast Cancer Cells.
Cytotoxicity of the Urokinase-Plasminogen Activator Inhibitor Carbamimidothioic Acid (4-Boronophenyl) Methyl Ester Hydrobromide (BC-11) on Triple-Negative MDA-MB231 Breast Cancer Cells.
Molecules (Basel, Switzerland) (2015-06-02)
Alessandra Longo, Mariangela Librizzi, Irina S Chuckowree, Christine B Baltus, John Spencer, Claudio Luparello
ABSTRACT
BC-11 is an easily synthesized simple thiouronium-substituted phenylboronic acid, which has been shown to be cytotoxic on triple negative MDA-MB231 breast cancer cells by inducing a perturbation of cell cycle when administered at a concentration equal to its ED50 at 72 h (117 μM). Exposure of cells to BC-11, either pre-absorbed with a soluble preparation of the N-terminal fragment of urokinase-plasminogen activator (uPa), or in co-treatment with two different EGFR inhibitors, indicated that: (i) BC-11 acts via binding to the N-terminus of the enzyme where uPa- and EGF receptor-recognizing sites are present, thereby abrogating the growth-sustaining effect resulting from receptor binding; and (ii) the co-presence of the EGFR inhibitor PD153035 potentiates BC-11's cytotoxicity. Exposure of cells to a higher concentration of BC-11 corresponding to its ED75 at 72 h (250 μM) caused additional impairment of mitochondrial activity, the production of reactive oxygen species and promotion of apoptosis. Therefore, BC-11 treatment appears to show potential for the development of this class of compounds in the prevention and/or therapy of "aggressive" breast carcinoma.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%