MilliporeSigma
  • Tenofovir monotherapy for hepatitis B after 1 year does not produce renal dysfunction, but is associated with hyperparathyroidism not related to vitamin D.
Tenofovir monotherapy for hepatitis B after 1 year does not produce renal dysfunction, but is associated with hyperparathyroidism not related to vitamin D.
European journal of gastroenterology & hepatology (2015-11-07)
Jose A Patricio, Patricia F Lopes, Thalia Medeiros, Guilherme F Mendes, Andrea A Silva, Eliane B Esberard, Jocemir R Lugon, Jorge R Almeida
ABSTRACT
Viral hepatitis B (VHB) represents a major public health problem. Studies from HIV multidrug patients have associated the use of tenofovir disoproxil fumarate (TDF) with renal dysfunction and phosphate wasting. The aim of this study was to examine the effect of year-long TDF monotherapy on renal function in VHB patients. We evaluated adult patients diagnosed with VHB before treatment initiation (T0), and after 3 and 12 months (T3 and T12) of TDF initiation. Estimated glomerular filtration rate (eGFR) was estimated by serum cystatin C and creatinine. In addition, urinary electrolytes and tubular biomarkers (cystatin C, β2-microglobulin and neutrophil gelatinase-associated lipocalin) were analyzed, as well as parathyroid hormone (PTH) and 25(OH)vitamin D levels. After 1 year, 32 patients completed the study, 22 (68.7%) men and 12 (37.5%) Whites, mean age 44.1±12.0 years. We found that serum electrolytes were similar at baseline and 3 or 12 months after initiation of TDF monotherapy. In addition, urinary fractional excretions of electrolytes as well as proteinuria, albuminuria, urinary β2-microglobulin, and urinary cystatin C showed no significant differences across the treatment timeline. There were also no statistical differences in the eGFR. There was a statistically significant increase in the PTH (Friedman's test, P=0.012), but the 25(OH)vitamin D levels were in the normal range in the beginning and did not change at the follow-up. Moreover, there was no correlation between the initial levels of vitamin D and the corresponding increases in the PTH values. If used as monotherapy in hepatitis B patients for a 12-month period, TDF is not associated with changes in either eGFR or a panel of urinary biomarkers. Serum and urinary electrolytes also remained unchanged. Of note, a significant increase in the PTH was found, although not related to the 25(OH)vitamin D initial status.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis