MilliporeSigma
  • A comparative study of ICH validated novel spectrophotometric techniques for resolving completely overlapping spectra of quaternary mixtures.
A comparative study of ICH validated novel spectrophotometric techniques for resolving completely overlapping spectra of quaternary mixtures.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-11-01)
Nouruddin W Ali, Nada S Abdelwahab, M Abdelkawy, Aml A Emam
ABSTRACT
A pharmaceutically marketed mixture of Yohimbine, Alpha-tocopheryl acetate, Niacin, and Caffeine co-formulated as a promising therapy for erectile dysfunction. Simultaneous determination of the aforementioned pharmaceutical formulation without prior separation steps was applied using mean centering of ratio spectra and triple divisor spectrophotometric methods. Mean centering of ratio spectra method depended on using the mean centered ratio spectra in three successive steps which eliminated the derivative steps and so the signal to noise ratio was improved. The absorption spectra of the prepared solutions were measured in the wavelength range of 215-300 nm in the concentration ranges of 1-15, 3-15, 1-20, and 3-15 μg mL(-1) for Yohimbine, Alpha-tocopheryl acetate, Niacin, and Caffeine, respectively. The amplitudes of the mean centered third ratio spectra were measured at 250 nm and 268 nm for Yohimbine and Alpha-tocopheryl acetate, respectively and at peak to peak 272-273 and 262-263 nm for Niacin and Caffeine, respectively. In triple divisor method each drug in the quaternary mixture was determined by dividing the spectrum of the quaternary mixture by a standard spectrum of a mixture containing equal concentrations of the other three drugs. First derivative of these ratio spectra was obtained where determination could be achieved without any interference from the other three drugs. Amplitudes of 1-15, 3-15, 1-15, and 3-15 μg mL(-1) were used for selective determination of Yohimbine, Alpha-tocopheryl acetate, Niacin, and Caffeine, respectively. Laboratory prepared mixtures were analyzed by the developed novel methods to investigate their selectivity also, Super Act® capsules were successfully analyzed to ensure absence of interference from additives. The developed methods were validated according to the ICH guidelines. The proposed methods were statistically compared with each other and with the reported methods; using student t-test, F-test, and one way ANOVA, where no significant difference was found with respect to accuracy and precision.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous