MilliporeSigma
  • Regeneration mechanisms of high-lithium content zirconates as CO2 capture sorbents: experimental measurements and theoretical investigations.
Regeneration mechanisms of high-lithium content zirconates as CO2 capture sorbents: experimental measurements and theoretical investigations.
Physical chemistry chemical physics : PCCP (2015-08-19)
Yuhua Duan, Jonathan Lekse
ABSTRACT
By combining TGA and XRD measurements with theoretical calculations of the capture of CO2 by lithium-rich zirconates (Li8ZrO6 and Li6Zr2O7), it has been demonstrated that the primary regeneration product during absorption/desorption cycling is in the form of Li2ZrO3. During absorption/desorption cycles, lithium-rich zirconates will be consumed and will not be regenerated. This result indicates that among known lithium zirconates, Li2ZrO3 is the best sorbent for CO2 capture.
Sigma-Aldrich
Lithium carbonate, 99.997% trace metals basis
Sigma-Aldrich
Lithium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Lithium-7Li2 carbonate, ≥99 atom % 7Li, 99% (CP)