MilliporeSigma
  • Selective cytotoxicity and cell death induced by human amniotic membrane in hepatocellular carcinoma.
Selective cytotoxicity and cell death induced by human amniotic membrane in hepatocellular carcinoma.
Medical oncology (Northwood, London, England) (2015-10-29)
A C Mamede, S Guerra, M Laranjo, M J Carvalho, R C Oliveira, A C Gonçalves, R Alves, L Prado Castro, A B Sarmento-Ribeiro, P Moura, A M Abrantes, C J Maia, M F Botelho
ABSTRACT
Hepatocellular carcinoma (HCC) has a worldwide high incidence and mortality. For this reason, it is essential to invest in new therapies for this type of cancer. Our team already proved that human amniotic membrane (hAM) is able to inhibit the metabolic activity of several human cancer cell lines, including HCC cell lines. Taking into account the previously performed work, this experimental study aimed to investigate the pathways by which hAM protein extracts (hAMPEs) act on HCC. Our results showed that hAMPE reduce the metabolic activity, protein content and DNA content in a dose- and time-dependent manner in all HCC cell lines. This therapy presents selective cytotoxicity, since it was not able to inhibit a non-tumorigenic human cell line. In addition, hAMPE induced cell morphology alterations in all HCC cell lines, but death type is cell line dependent, as proved by in vitro and in vivo studies. In conclusion, hAMPE have a promising role in HCC therapy, since it is capable of inducing HCC cytotoxicity and cell death.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture