MilliporeSigma
  • Dot-ELISA Rapid Test Using Recombinant 56-kDa Protein Antigens for Serodiagnosis of Scrub Typhus.
Dot-ELISA Rapid Test Using Recombinant 56-kDa Protein Antigens for Serodiagnosis of Scrub Typhus.
The American journal of tropical medicine and hygiene (2015-03-25)
Wuttikon Rodkvamtook, Zhiwen Zhang, Chien-Chung Chao, Erin Huber, Dharadhida Bodhidatta, Jariyanart Gaywee, John Grieco, Narongrid Sirisopana, Manerat Kityapan, Michael Lewis, Wei-Mei Ching
ABSTRACT
We developed a rapid dot-enzyme-linked immunosorbent assay (dot-ELISA) using the combination of recombinant 56-kDa protein antigens that exhibited broad reactivity with serum antibodies against the four most prevalent strains (Karp, Kato, Gilliam, and TA763) of Orientia tsutsugamushi. The assay is rapid (30 minutes), and can be done at room temperature, and results can be read by the naked eye. Only a simple shaker is required to wash the membrane. Sera from 338 patients suspected of being ill with scrub typhus from rural hospitals around Thailand were tested using this dot-ELISA. Seventy-five (22.2%) patients were found to be positive. The sensitivity and specificity of dot-ELISA were determined using the indirect immunofluorescent assay (IFA) test as the gold standard, with the cutoff titer of immunoglobulin peroxidase conjugate M (IgM)/G (IgG) greater than 1:400/1:400. The dot-ELISA had a sensitivity of 98.5%, a specificity of 96.3%, a positive predictive value of 86.7%, and a negative predictive value of 99.6% for the acute-phase specimens. The results indicate that dot-ELISA rapid test using recombinant 56-kDa protein antigen was comparable with the IFA test and may be very useful for the diagnosis of scrub typhus in rural hospitals, where IFA is not available.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Triton X-114
Supelco
IGEPAL® CA-630
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Triton X-405 solution, 70% in H2O
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)