MilliporeSigma
A synthetic peptide adhesion epitope as a novel antimicrobial agent.
Nature biotechnology (1999-01-27)
C G Kelly, J S Younson, B Y Hikmat, S M Todryk, M Czisch, P I Haris, I R Flindall, C Newby, A I Mallet, J K Ma, T Lehner
ABSTRACT
The earliest step in microbial infection is adherence by specific microbial adhesins to the mucosa of the oro-intestinal, nasorespiratory, or genitourinary tract. We inhibited binding of a cell surface adhesin of Streptococcus mutans to salivary receptors in vitro, as measured by surface plasmon resonance, using a synthetic peptide (p1025) corresponding to residues 1025-1044 of the adhesin. Two residues within p1025 that contribute to binding (Q1025, E1037) were identified by site-directed mutagenesis. In an in vivo human streptococcal adhesion model, direct application of p1025 to the teeth prevented recolonization of S. mutans but not Actinomyces, as compared with a control peptide or saline. This novel antimicrobial strategy, applying competitive peptide inhibitors of adhesion, may be used against other microorganisms in which adhesins mediate colonization of mucosal surfaces.