MilliporeSigma
  • Oncolytic virus carrying shRNA targeting SATB1 inhibits prostate cancer growth and metastasis.
Oncolytic virus carrying shRNA targeting SATB1 inhibits prostate cancer growth and metastasis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-06-19)
Li-jun Mao, Jie Zhang, Ning Liu, Li Fan, Dong-rong Yang, Bo-xin Xue, Yu-xi Shan, Jun-nian Zheng
ABSTRACT
Recent studies suggest that SATB1 is a promising therapeutic target for prostate cancer. To develop novel SATB1-based therapeutic agents for prostate cancer, in this study, we aimed to construct ZD55-SATB1, an oncolytic adenovirus ZD55 carrying shRNA targeting SATB1, and investigate its effects on the inhibition of prostate cancer growth and metastasis. ZD55-SATB1 was constructed and used to infect human prostate cancer cell lines DU145 and LNCaP. The inhibitory effect of ZD55-SATB1 on SATB1 expression was evaluated by reverse transcription polymerase chain reaction (RT-PCR) and Western blot analysis. The cytotoxicity of ZD55-SATB1 was detected by MTT assay. Cell invasion was detected by Matrigel invasion assay. The in vivo antitumor activities of ZD55-SATB1 were evaluated in xenograft mouse model. We found that ZD55-SATB1 selectively replicated and significantly reduced SATB1 expression in DU145 and LNCaP cells. ZD55-SATB1 effectively inhibited the viability and invasion of DU145 and LNCaP cells in vitro and inhibited prostate cancer growth and metastasis in xenograft nude mice. In conclusion, replicative oncolytic adenovirus armed with SATB1 shRNA exhibits effective antitumor effect in human prostate cancer. Our study provides the basis for the development of ZD55-SATB1 for the treatment of prostate cancer.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M