MilliporeSigma
  • Down-regulated expression of miR-134 contributes to paclitaxel resistance in human ovarian cancer cells.
Down-regulated expression of miR-134 contributes to paclitaxel resistance in human ovarian cancer cells.
FEBS letters (2015-09-13)
Ting Shuang, Min Wang, Cong Shi, Yingying Zhou, Dandan Wang
ABSTRACT
MiR-134 has been reported to have a role in the development and progression of various cancers. In this study, we found that miR-134 expression was significantly decreased in chemo-resistant serous epithelial ovarian cancer (EOC) patients. Over-expression of miR-134 enhanced the sensitivity of SKOV3-TR30 cells to paclitaxel, and increased paclitaxel-induced apoptosis. Further, Pak2 was identified as a direct target of miR-134, and Pak2-specific siRNA increased cell inhibition rate and promoted paclitaxal-induced apoptosis. By regulating Pak2 expression, miR-134 could mediate Bad phosphorylation at Ser112 and Ser136, which affected cell survival and apoptosis. In conclusion, our findings indicate that repression of miR-134 and consequent up-regulation of Pak2 might contribute to paclitaxel resistance.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pak2
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
MISSION® esiRNA, targeting human PAK2
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)