MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Magnetic field dependent electro-conductivity of the graphite doped magnetorheological plastomers.
Magnetic field dependent electro-conductivity of the graphite doped magnetorheological plastomers.
Soft matter (2015-08-04)
Haoming Pang, Shouhu Xuan, Taixiang Liu, Xinglong Gong
ABSTRACT
In this work we reported a novel graphite doped conductive magnetorheological plastomer (GMRP) with magnetic field dependent electro-conductivity. The conductivity of the GMRPs increased by increasing the content of the graphite particles, while it decreased with the graphite size. When the graphite content reached 15 wt%, the conductivity of GMRPs is approximately 10 000 times higher than the non-doped MRP. Because the iron particles in the GMRPs were magnetic, the conductivity of the GMRPs was magnetically sensitive. Upon applying a 780 mT magnetic field, the electric conductivity could increase about 1000 times larger than the one under zero magnetic field. A particle-particle resistance model was developed to investigate the influence of the magnetic field and graphite doping on the conductivity, and the fitting curve matched the experimental results very well. Finally, a magnetically controllable on-off switch based on GMRPs was proposed and its working mechanism was discussed.
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, ≥99%
Sigma-Aldrich
Tolylene-2,4-diisocyanate, technical grade, 80%
Sigma-Aldrich
Tolylene-2,6-diisocyanate, 97%