MilliporeSigma
  • Understanding the lateral movement of particles adsorbed at a solid-liquid interface.
Understanding the lateral movement of particles adsorbed at a solid-liquid interface.
Journal of colloid and interface science (2015-05-26)
Kunal Savaji, Xue Li, Alexander Couzis
ABSTRACT
In this paper we study the phenomenon of lateral movement of particles that are electrostatically adsorbed at a solid-liquid interface. The experimental system involves negatively charged silica particles of two different sizes (65 nm and 90 nm) that are exposed to the positively charged solid surface (silane coated silicon wafer) in sequential steps. The particle-adsorbed wafers are analyzed under a scanning electron microscope and the images are processed to determine the pair-correlation function for the particles adsorbed in the first step. From the pair correlation data and the particle surface coverage data we show that the adsorbed particles are mobile at the solid-liquid interface. In specific, we show that the adsorbed particles are mobile at the solid-liquid interface when there is a driving force for the adsorbed particles to move. The driving force in the scheme of experiments discussed in this paper is the reduction in the free energy of the system.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
CAPS, BioXtra, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
CAPS, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%