MilliporeSigma
  • Mechanisms of growth-promoting and tumor-protecting effects of epithelial nicotinic acetylcholine receptors.
Mechanisms of growth-promoting and tumor-protecting effects of epithelial nicotinic acetylcholine receptors.
International immunopharmacology (2015-06-14)
Alex I Chernyavsky, Igor B Shchepotin, Sergei A Grando
ABSTRACT
Although the role of nicotine as a carcinogen is debatable, it is widely accepted that it contributes to cancer by promoting growth and survival of mutated cell clones and protecting them from the chemo- and radiotherapy-induced apoptosis. On the cell membrane (cm), the nicotinic acetylcholine (ACh) receptors (nAChRs) implement upregulation of proliferative and survival genes. Nicotine also can permeate cells and activate mitochondrial (mt)-nAChRs coupled to inhibition of the mitochondrial permeability transition pore (mPTP) opening, thus preventing apoptosis. In this study, we sought to pin down principal mechanisms mediating the tumor-promoting activities of nicotine resulting from activation of cm- and mt-nAChRs in oral and lung cancer cells, SCC25 and SW900, respectively. Activated cm-nAChRs were found to form complexes with receptors for EGF and VEGEF via the α7 and β2 nAChR subunits, respectively, whereas activated mt-nAChRs physically associated with the intramitochondrial protein kinases PI3K and Src via the α7 and β4 subunits. This was associated with upregulated expression of cyclin D1/activation of ERK1/2 and inhibition of mPTP opening, respectively, as well as upregulated proliferation and resistance to H(2)O(2)-induced apoptosis. The molecular synergy between cm-nAChRs and growth factor receptors helps explain how one biological mediator, such as ACh, can modulate activity of the other, such as a growth factor, and vice versa. Establishment of functional coupling of mt-nAChRs to regulation of mPTP opening provides a novel mechanism of nicotine-dependent protection from cell death. Further elucidation of this novel mechanism of tumor-promoting activities of nicotine should have a strong translational impact, because extraneuronal nAChRs may provide a novel molecular target to prevent, reverse, or retard progression of both nicotine-related and unrelated cancers.
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Hemicholinium-3, solid, ≥95% (HPLC)
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
EGF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid