MilliporeSigma
  • Gastrointestinal immunity against tryptophan hydroxylase-1, aromatic L-amino-acid decarboxylase, AIE-75, villin and Paneth cells in APECED.
Gastrointestinal immunity against tryptophan hydroxylase-1, aromatic L-amino-acid decarboxylase, AIE-75, villin and Paneth cells in APECED.
Clinical immunology (Orlando, Fla.) (2015-03-26)
Nicolas Kluger, Martta Jokinen, Anu Lintulahti, Kai Krohn, Annamari Ranki
ABSTRACT
Gastrointestinal dysfunction is a disabling manifestation of APECED possibly related to an autoimmune intestinal aggression. We evaluated its features in a cohort of 31 Finnish patients. The most frequent manifestations were constipation (48%), diarrhea, dysphagia and retrosternal pain (45%). AADC and TPH-1 autoantibodies were detected in 51% and 45% of the patients, respectively. Forty-three percent displayed a T-cell response to AADC. One third of the patients also had AIE-75 (33%) and villin (29%)-specific autoantibodies while antibodies against brush borders and Paneth cells were detected in 29% and 20%, respectively. Intestinal IL-17 expression was absent/decreased in 77% of the cases. Duodenal CgA and serotonin expression was absent/decreased in 50% and 66% of the patients, respectively. Constipation correlated with lacking serotonin expression and AADC antibodies (p < 0.05).
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder