MilliporeSigma
  • Dysregulation of zinc finger protein, X-linked (ZFX) impairs cell proliferation and induces apoptosis in human oral squamous cell carcinorma.
Dysregulation of zinc finger protein, X-linked (ZFX) impairs cell proliferation and induces apoptosis in human oral squamous cell carcinorma.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-04-29)
Hongzhi Ma, Fan Yang, Meng Lian, Ru Wang, Haizhou Wang, Ling Feng, Qian Shi, Jugao Fang
ABSTRACT
Zinc finger protein, X-linked (ZFX) is a transcriptional factor involved in many physiological processes such as embryonic stem cell survival and self-renewal. Though ZFX dysfunctions have been identified in variant human diseases and especially in cancers, its pathological roles have not been fully addressed. Here, we explored the relationship between ZFX expression and squamous cell carcinoma (SCC) of the tongue. We found that ZFX expression was significantly higher in tongue SCC tumors as compared to tumor-adjacent normal tissues. Furthermore, ZFX knockdown impeded cell proliferation, impaired colony formation ability, and lead to cell cycle arrest while induced cell apoptosis in human tongue squamous cell carcinoma cell line Tca-8113. Our results provide evidence suggesting that ZFX overexpression is associated with the development of tongue SCC and ZFX knockdown is a potential treatment for tumor suppression.
Product Description
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests