MilliporeSigma
  • Incipient cytotoxicity: A time-independent measure of cytotoxic potency in vitro.
Incipient cytotoxicity: A time-independent measure of cytotoxic potency in vitro.
Toxicology (2015-07-15)
Michael Gülden, Daria Kähler, Hasso Seibert
ABSTRACT
Time is an important determinant of toxicity but largely ignored in in vitro toxicity assays where exposure times chosen are rather arbitrary. To investigate the impact of time on the cytotoxic potency of chemicals in vitro, the concentration dependent cytotoxic action of selected chemicals (surfactants, metals, oxidative stressors, a mitochondrial poison) was determined after various exposure times (1-72 h) in cultures of Balb/c 3T3 cells. Time affected the cytotoxic potency as well as the cytotoxic efficacy. The median cytotoxic concentrations, EC50, decreased and in most cases approached an "incipient" value, EC50,∞, within 72 h. Cytotoxicity due to mitochondrial insult occurred after a threshold time which was dependent on the medium glucose concentration. Within the chemicals studied the extent of potency change with time ranged from 3- to >1000-fold and the "time to incipient cytotoxicity", tic, from 4 to >72 h. Hence, also the relative cytotoxic potencies depend on exposure time. Ignoring this may lead to severe bias in toxicological hazard and risk assessment. Therefore it is recommended to determine the incipient cytotoxic potency of chemical compounds, represented by, e.g., the incipient median effect (EC50,∞), no effect (NEC∞) or lowest effect concentrations (LEC∞) instead of measures obtained after arbitrary exposure times. If this is not possible, the 72 h-potency measurements appear to be useful surrogates. These time-independent incipient potency values can be reasonably compared between substances, endpoints, cells and biological test systems and may serve to define points of departure for quantitative in vitro-in vivo extrapolations.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Cumene, 98%
Sigma-Aldrich
cis-4-Hydroxy-L-proline, collagen synthesis inhibitor