MilliporeSigma
Altered mitochondrial response to activation of T-cells in neonate.
Acta physiologica Hungarica (2015-06-24)
G Mészáros, Cs Orbán, A Kaposi, G Toldi, B Gyarmati, T Tulassay, Barna Vásárhelyi
ABSTRACT
Mitochondrial functions have a major impact on T-cell functionality. In this study we characterized whether mitochondrial function in the neonatal T-cells differs from that in the adult T-cells during short T-cell activation. We used flow cytometry methods to test mitochondrial mass and to monitor mitochondrial Ca²⁺ levels, mitochondrial potential and superoxide generation in parallel with cytoplasmic Ca²⁺ levels during phythohaemagglutinine-induced activation of CD4+ and CD8+ T-cells of 12 term neonates and 11 healthy adults. Baseline mitochondrial mass of CD4+ and CD8+ cells was lower in the neonate than in the adult. In comparison with the adult, neonatal resting CD4+ T-cells had lower cytoplasmic Ca²⁺ levels and this was associated with normal activation induced Ca²⁺-response. During short-term activation cytoplasmic Ca²⁺-response was lower in neonatal than in adult CD8+ T-cells. Mitochondrial Ca²⁺ uptake was increased in CD4+ neonatal T cells while it decreased in CD8+ T-cells. Mitochondrial depolarization was increased in CD4+ and decreased in CD8+ neonatal T-cells compared to adults. Superoxide generation was higher and equal in neonatal CD4+ and CD8+ cells, respectively, compared to the adult ones. Our data suggest that neonatal T-cells exhibit marked differences in mitochondrial function and superoxide generation compared to adult T-cells.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%