MilliporeSigma
  • MiR-361-5p inhibits colorectal and gastric cancer growth and metastasis by targeting staphylococcal nuclease domain containing-1.
MiR-361-5p inhibits colorectal and gastric cancer growth and metastasis by targeting staphylococcal nuclease domain containing-1.
Oncotarget (2015-05-13)
Fei Ma, Hongjiang Song, Baoliang Guo, Yuxin Zhang, Yasheng Zheng, Chengchun Lin, Ying Wu, Guijie Guan, Ruihua Sha, Qingxin Zhou, Dejun Wang, Xinglu Zhou, Juan Li, Xiaohui Qiu
ABSTRACT
MicroRNAs (miRs) function as key regulators of gene expression and their deregulation is associated with the carcinogenesis of various cancers. In the present study, we investigated the biological role and mechanism of miR-361-5p in colorectal carcinoma (CRC) and gastric cancer (GC). We showed that microRNA-361-5p (miR-361-5p) was down-regulated in CRC and GC in comparison to the controls. Meanwhile, the expression levels of miR-361-5p negatively correlated with lung metastasis and prognosis in clinical CRC patients. Overexpression of miR-361-5p markedly suppressed proliferation, migration and invasion of cancer cells. Additionally, this phenotype could be partially rescued by the ectopic expression of staphylococcal nuclease domain containing-1 (SND1). SND1 was identified as a target of miR-361-5p using bioinformatics analysis and in vitro luciferase reporter assays. In turn, SND1 bound to pre-miR-361-5p and suppressed the expression of miR-361-5p, thus exerting a feedback loop. Most interestingly, in vivo studies showed that restoration of miR-361-5p significantly inhibited tumor growth and especially the lung metastasis in nude mice. Therefore, it could be concluded that miR-361-5p functions as a tumor-suppressive miRNA through directly binding to SND1, highlighting its potential as a novel agent for the treatment of patients with CRC and GC.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Snd1
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting human SND1