MilliporeSigma
Concentrated assemblies of magnetic nanoparticles in ionic liquids.
Faraday discussions (2015-05-12)
Marianna Mamusa, Juliette Sirieix-Plénet, Régine Perzynski, Fabrice Cousin, Emmanuelle Dubois, Véronique Peyre
ABSTRACT
Maghemite (γ-Fe2O3) nanoparticles (NPs) can be successfully dispersed in a protic ionic liquid, ethylammonium nitrate (EAN), by transfer from aqueous dispersions into EAN. As the aqueous systems are well controlled, several parameters can be tuned. Their crucial role towards the interparticle potential and the structure of the dispersions is evidenced: (i) the size of the NPs tunes the interparticle attraction monitoring dispersions to be either monophasic or gas-liquid-like phase separated; (ii) the nature of the initial counterion in water (here sodium, lithium or ethylammonium) and the amount of added water (<20 vol%) modulate the interparticle repulsion. Very concentrated dispersions with a volume fraction of around 25% are obtained thanks to the gas-liquid-like phase separations. Such conclusions are derived from a fine structural and dynamical study of the dispersions on a large range of spatial scales by coupling several techniques: chemical analyses, optical microscopy, dynamic light scattering, magneto-optic birefringence and small angle scattering.
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis