MilliporeSigma
  • Noninvasive Biomarker Candidates for Cadmium-Induced Nephrotoxicity by 2DE/MALDI-TOF-MS and SILAC/LC-MS Proteomic Analyses.
Noninvasive Biomarker Candidates for Cadmium-Induced Nephrotoxicity by 2DE/MALDI-TOF-MS and SILAC/LC-MS Proteomic Analyses.
Toxicological sciences : an official journal of the Society of Toxicology (2015-08-12)
Sun Young Kim, Hye Min Lee, Kyeong Seok Kim, Hyung Sik Kim, Aree Moon
ABSTRACT
Cadmium (Cd(2+)) is a major environmental pollutant that induces cytotoxicity by heavy-metal accumulation. Prolonged Cd(2+) exposure leads to cell damage by oxidative stress mainly in the kidneys, a critical organ for detoxification. To identify reliable on invasive protein biomarkers for Cd(2+)-induced nephrotoxicity, we performed 2-dimensional gel electrophoresis/matrix-assisted laser desorption/ionization time of flight mass spectra and stable isotope labeling by amino acids in cell culture/liquid chromatography-mass spectrometry analyses using conditioned media (CM) of HK-2 human kidney epithelial cells treated with CdCl2. Here, we identified heat shock cognate 71 kDa protein isoform1 (HSPA8) and α-enolase (ENO1) as potential biomarker candidates for the evaluation of Cd(2+)-induced nephrotoxicity. Treatment with CdCl2 increased the protein level of HSPA8 in CM and lysates of HK-2 cells. The mRNA level of HSPA8 was also increased by CdCl2 treatment, indicating transcriptional regulation. The level of ENO1 was increased in CM, but not in lysates of CdCl2-treated HK-2 cells. CdCl2 did not affect the mRNA level of ENO1. We provide evidence that the increases of HSPA8 and ENO1 in CM were due to Cd(2+)-induced cell death through oxidative stress. The increases of HSPA8 and ENO1 levels were also detected in CM of HK-2 cells treated with other nephrotoxic agents, such as HgCl2, NaAsO2, cisplatin, amphotericin B, and cyclosporine A. Urine and kidney tissues of CdCl2-treated rats showed increased levels of HSPA8. Taken together, this study identified HSPA8 and ENO1 as noninvasive biomarker candidates by 2 comparative proteomic analyses. These new biomarker candidates may have potential as alternatives to traditional biomarkers for the efficient and sensitive assessment of nephrotoxicity.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, puriss., suitable for, meets analytical specification of Ph. Eur., BP, USP,FCC
Sigma-Aldrich
Copper(II) sulfate pentahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%
Sigma-Aldrich
Mercury(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Mercury(II) chloride, ACS reagent, ≥99.5%
Sigma-Aldrich
D-Glyceraldehyde 3-phosphate solution, 8-13 mg/mL in H2O
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
(±)-CPP, solid
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder