MilliporeSigma
  • Study of the BCG Vaccine-Induced Cellular Immune Response in Schoolchildren in Antananarivo, Madagascar.
Study of the BCG Vaccine-Induced Cellular Immune Response in Schoolchildren in Antananarivo, Madagascar.
PloS one (2015-07-28)
Paulo Ranaivomanana, Vaomalala Raharimanga, Patrice M Dubois, Vincent Richard, Voahangy Rasolofo Razanamparany
ABSTRACT
Although the Bacillus Calmette-Guérin vaccine (BCG) protects young children against serious forms of TB, protection against pulmonary TB is variable. We assessed BCG vaccine-induced cellular immune responses and determined for how long they could be detected during childhood in Antananarivo, Madagascar. We assessed BCG vaccine-induced cellular immune responses by TST and IGRA (in-house ELISPOT assay) using BCG and PPD as stimulation antigen, and compared results between vaccinated and non-vaccinated schoolchildren of two age groups, 6-7 and 13-14 years old. Three hundred and sixty-three healthy schoolchildren were enrolled. TST was performed on 351 children and IGRA on 142. A high proportion (66%; 229/343) of the children had no TST reactivity (induration size 0 mm). TST-positive responses (≥15 mm) were more prevalent among 13-14 year-old (31.7%) than 6-7 year old (16.5%) children, both in the non-vaccinated (43% vs. 9%, p<0.001) and vaccinated (29% vs. 13%, p=0.002) subgroups. There were no significant differences in TST responses between vaccinated and non-vaccinated children in either of the age groups. The IGRA response to BCG and to PPD stimulation was not significantly different according to BCG vaccination record or to age group. A high rate (15.5%; 22/142) of indeterminate IGRA responses was observed. There was very poor agreement between TST and IGRA-PPD findings (k= 0.08) and between TST and IGRA-BCG findings (k= 0.02). Analysis of TST and IGRA response to stimulation with BCG and PPD revealed no difference in immune response between BCG-vaccinated and non-vaccinated children; also no decrease of the BCG vaccine-induced cellular immune response over time was observed. We conclude that TST and IGRA have limitations in assessing a role of BCG or tuberculosis-related immunity.
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Leucoagglutinin PHA-L, lyophilized powder
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%