MilliporeSigma
  • Resveratrol decreases the insoluble Aβ1-42 level in hippocampus and protects the integrity of the blood-brain barrier in AD rats.
Resveratrol decreases the insoluble Aβ1-42 level in hippocampus and protects the integrity of the blood-brain barrier in AD rats.
Neuroscience (2015-10-11)
H F Zhao, N Li, Q Wang, X J Cheng, X M Li, T T Liu
ABSTRACT
Our previous studies demonstrated resveratrol (Res) administration protected Alzheimer's disease (AD) rats from developing memory decline by anti-oxidation. Beta-amyloid peptide 1-42 (Aβ1-42) is not only the primary protein component of senile plaques in AD but also is believed to play an important part in its pathology. Increasing evidence has shown neuroinflammation and the integrity of the blood-brain barrier (BBB) is closely related to the pathogenesis of AD. The aim of the present study is to further elucidate whether Res prevents AD rats from inflammation induced by Aβ1-42 and protects the integrity of BBB. Rats were divided into six groups: (1) ovariectomized (OVX)+D-galactose (D-gal) 100mg/kg group (OVX+D-gal); (2-4) OVX, D-gal and Res 20, 40 and 80 mg/kg treated groups; and (5) OVX, D-gal and estradiol valerate 0.8 mg/kg treated group (ET); (6) Sham control group. 12 weeks later, Res 40 and 80 mg/kg treatment exhibited a significant decrease of Aβ1-42 compared with the OVX+D-gal rats of hippocampus, which was accompanied by decreased expression of advanced glycation endproducts (RAGE), matrix metalloprotein-9 (MMP-9), nuclear factor kappaB (NF-κB) and the increase of Claudin-5. These results suggest that Res is useful not only in protecting OVX+D-gal rats from neuroinflammation mediated by Aβ1-42 by decreasing the expression of NF-κB but also the integrity of BBB by increasing Claudin-5 and decreasing RAGE, MMP-9.
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture