MilliporeSigma
  • The usability of a 15-gene hypoxia classifier as a universal hypoxia profile in various cancer cell types.
The usability of a 15-gene hypoxia classifier as a universal hypoxia profile in various cancer cell types.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology (2015-07-15)
Brita Singers Sørensen, Anders Knudsen, Catja Foged Wittrup, Steffen Nielsen, Ninna Aggerholm-Pedersen, Morten Busk, Michael Horsman, Morten Høyer, Pierre Nourdine Bouchelouche, Jens Overgaard, Jan Alsner
ABSTRACT
A 15-gene hypoxia profile has previously demonstrated to have both prognostic and predictive impact for hypoxic modification in squamous cell carcinoma of the head and neck. This gene expression profile may also have a prognostic value in other histological cancer types, and could potentially have a function as a universal hypoxia profile. The hypoxia induced upregulation of the included genes, and the validity of the previously used reference genes was established in this study, in a range of different cell lines representing carcinomas of the prostate, colon, and esophagus. Eleven adenocarcinoma and one squamous cell lines: Six colon carcinomas (HTC8, HT29, LS174T, SW116, SW948 and T48), 3 prostate carcinomas (LNCaP, DU-145 and PC-3) and 3 esophagus carcinoma cell lines (OE19, OE21 and OE33) were cultured under normoxic or hypoxic conditions (0% O2) for 24hours. Total RNA was extracted and gene expression levels measured by qPCR. For each tissue type, individual reference genes were selected and applied in the normalization of the relative expression levels. In all three tissue types, individual, optimal, reference genes were selected. In the analysis of the hypoxia induced genes, both the original reference genes and the new selected reference genes were used. There was no significant difference in the obtained data. The gene expression analysis demonstrated cell line specific differences in the hypoxia response of the 15 genes, with BNIP3 not being upregulated at hypoxic conditions in 3 out of 6 colon cancer cell lines, and ALDOA in OE21 and FAM162A and SLC2A1 in SW116 only showing limited hypoxia induction. Furthermore, in the esophagus cell lines, the normoxic and hypoxic expression levels of LOX and BNIP3 were below the detection limit in OE19 and OE33, respectively. However, a combined analysis of the 15 genes in both adenocarcinoma cell lines and squamous carcinoma cell lines demonstrated a very consistent expression pattern in hypoxic induced gene expression across all cell lines. This study addressed the tissue type dependency of hypoxia induced genes included in a 15-gene hypoxic profile in carcinoma cell lines from prostate, colon, and esophagus cancer, and demonstrated that in vitro, with minor fluctuations, the genes in the hypoxic profile are hypoxia inducible, and the hypoxia profile may be applicable in other sites than HNSCC.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications