MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Determination of selected veterinary antimicrobials in poultry excreta by UHPLC-MS/MS, for application in Salmonella control programs.
Determination of selected veterinary antimicrobials in poultry excreta by UHPLC-MS/MS, for application in Salmonella control programs.
Analytical and bioanalytical chemistry (2015-01-31)
Brecht Gorissen, Tim Reyns, Mathias Devreese, Patrick De Backer, Joris Van Loco, Siska Croubels
ABSTRACT
The most important source of Salmonella spp. infection in humans is by the consumption of contaminated poultry products. Due to the risk of resistance development and its transfer from animals to humans, the Belgian Royal Decree concerning the eradication of Salmonella (C-2007/22784) prohibits treatment of poultry with antimicrobials against zoonotic Salmonella spp. To uncover illicit use, an analytical method using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) for the determination of antimicrobial residues in poultry excreta was developed and validated for classes having an active spectrum against Salmonella spp. in poultry: β-lactams (amoxicillin and penicillin V), fluoroquinolones (enrofloxacin, difloxacin, and flumequine), polymyxins (colistin), sulfonamides in combination with trimethoprim (sulfachloropyridazine, sulfadiazine, and sulfaclozine), and tetracyclines (chlortetracycline and doxycycline). A generic and high-throughput sample preparation was developed. Extraction of samples was performed by ultrasonication using a combination of acetonitrile and McIlvaine buffer, followed by centrifugation and filtration prior to analysis. The method was validated according to Commission Decision 2002/657/EC for linearity, apparent recovery/trueness, repeatability, reproducibility, limit of quantification, limit of detection, specificity, matrix effect, and storage stability in matrix. To demonstrate the applicability of the method, an in vivo experiment was conducted. For each antimicrobial class, one registered drug was selected and administered in the drinking water to two laying hens. Excreta samples were collected every 12 h during and until 2 days after treatment and analyzed using the developed method.
Sigma-Aldrich
Chlortetracycline hydrochloride, BioReagent, suitable for fluorescence, from Streptomyces aureofaciens, ≥85.0% (HPLC)
Sigma-Aldrich
Chlortetracycline hydrochloride, ≥91.0% dry basis (HPLC)
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Demeclocycline hydrochloride, powder or crystals
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, Molecular Biology, ≥99.0% (T)
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)