Merck
  • Antigen targeting to dendritic cells combined with transient regulatory T cell inhibition results in long-term tumor regression.
Antigen targeting to dendritic cells combined with transient regulatory T cell inhibition results in long-term tumor regression.
Oncoimmunology (2015-09-26)
Wendy Wj Unger, Christian T Mayer, Steef Engels, Christina Hesse, Maurizio Perdicchio, Franz Puttur, Ingeborg Streng-Ouwehand, Manja Litjens, Hakan Kalay, Luciana Berod, Tim Sparwasser, Yvette van Kooyk
ABSTRACT
Therapeutic vaccinations against cancer are still largely ineffective. Major caveats are inefficient delivery of tumor antigens to dendritic cells (DCs) and excessive immune suppression by
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Ethidium bromide monoazide, ≥95% (HPLC), solid
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis