MilliporeSigma
  • Synthesis of copper ion incorporated horseradish peroxidase-based hybrid nanoflowers for enhanced catalytic activity and stability.
Synthesis of copper ion incorporated horseradish peroxidase-based hybrid nanoflowers for enhanced catalytic activity and stability.
Dalton transactions (Cambridge, England : 2003) (2015-05-06)
Burcu Somturk, Mehmet Hancer, Ismail Ocsoy, Nalan Özdemir
ABSTRACT
In this study, we report the preparation, catalytic activity and stability of a hybrid nanoflower (hNF) formed from horseradish peroxidase (HRP) enzyme and copper ions (Cu(2+)). We studied the morphology of hNFs as a function of the concentrations of copper (Cu(2+)) ions, chloride ions (Cl(-)) and HRP enzyme, the pH of the buffer solution (phosphate buffered saline), and the temperature of the reaction. The effects of morphology on the catalytic activity and stability of hNFs were evaluated by oxidation of guaiacol (2-methoxyphenol) to colored 3,3-dimethoxy-4,4-diphenoquinone in the presence of hydrogen peroxide (H2O2). The enhanced activity of hNFs synthesized (from 0.02 mg mL(-1) HRP in 10 mM PBS (pH 7.4) at +4 °C) was 17 595 U mg(-1), which was ∼300% higher than free HRP in PBS, where it achieved an activity of 5952 U mg(-1). In terms of stability, these hNFs stored in PBS buffer at +4 °C and room temperature (RT = 20 °C) lost 4% and 20%, respectively, of their initial catalytic activities within 30 days. Finally, we demonstrated that these hNFs can be utilized as sensors for the detection of dopamine.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Guaiacol, oxidation indicator