MilliporeSigma
  • Vitamin D deficiency contributes to the reduction and impaired function of naïve CD45RA⁺ regulatory T cell in chronic heart failure.
Vitamin D deficiency contributes to the reduction and impaired function of naïve CD45RA⁺ regulatory T cell in chronic heart failure.
Journal of immunology research (2015-05-20)
Yan-hui Ma, Yun-lan Zhou, Chao-yan Yue, Guang-hui Zhang, Lin Deng, Guo-hua Xie, Wei-ping Xu, Li-song Shen
ABSTRACT
The effect of vitamin D pertinent to cardiovascular health on the heart itself is considered to shift toward an anti-inflammatory response in chronic heart failure (CHF); however, its underlying mechanism is not completely understood. In this study, we demonstrated that plasma 25(OH)D level, negatively associated with NT-ProBNP, correlated with the decreased Treg in CHF compared to the patients with other cardiovascular diseases and healthy and older donors. Naïve Treg cell (CD4(+)CD45RA(+)Foxp3(lo)T) subset, rather than whole Treg cells, contributes to the reduction of Treg in CHF. 1,25(OH)2D treatment maintained partial expression of CD45RA on CD4(+)T cell after αCD3/CD28 monoclonal antibodies activation and ameliorated the impaired CD4(+)CD45RA(+)T cell function from CHF patients through upregulating Foxp3 expression and IL-10 secretion in vitro. Low level of vitamin D receptor (VDR) was detected in CD4(+)CD45RA(+)T cell of CHF than control, while 1,25(OH)2D treatment increased the VDR expression to exert its immunosuppression on T cell. The results of this study might provide tangible evidence to our knowledge of the impact of vitamin D supplementation on naïve Tregs, which may offer new means of preventing and treating CHF.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES