MilliporeSigma
  • Neural tissue regeneration in experimental brain injury model with channeled scaffolds of acrylate copolymers.
Neural tissue regeneration in experimental brain injury model with channeled scaffolds of acrylate copolymers.
Neuroscience letters (2015-05-20)
Cristina Martínez-Ramos, Ulises Gómez-Pinedo, Maria Angeles Garcia Esparza, José Miguel Soria, Juan A Barcia, Manuel Monleón Pradas
ABSTRACT
The objective of the present study was to evaluate the biocompatibility and cell hosting ability of a copolymer scaffold based on ethyl acrylate (EA) and hydroxyl ethyl acrylate (HEA) in vivo after an experimental brain injury. Wistar rats were subjected to cryogenic traumatic brain injury. We evaluated the tissue response to the implanted materials after 8 weeks. The materials were implanted devoid of cells; they provoked a minimal scar response by the host tissue and permitted the invasion of neurons and glia inside them. We also found new blood vessels surrounding and inside the implant. Thus, the copolymer scaffold proves to offer a suitable environment producing a cellular network potentially useful in brain repair after brain injury.
Sigma-Aldrich
2-Hydroxyethyl acrylate, 96%, contains 200-650 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), recrystallized from methanol, 99%
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized
Sigma-Aldrich
DAPI, for nucleic acid staining