MilliporeSigma
  • Investigation of Drug Release from PEO Tablet Matrices in the Presence of Vitamin E as Antioxidant.
Investigation of Drug Release from PEO Tablet Matrices in the Presence of Vitamin E as Antioxidant.
Current drug delivery (2015-03-27)
Saeed Shojaee, Ali Nokhodchi, Iain Cumming, Amjad Alhalaweh, Waseem Kaialy
ABSTRACT
The objective of this study was to investigate the influence of drug type on the release of drug from PEO matrix tablets accompanied with the impact of vitamin E succinate as antioxidant. The result showed that the presence of vitamin E promoted a stable release rate of soluble drug propranolol HCl from aged PEO matrix tablets, which was similar to fresh sample, regardless of molecular weight (MW) of PEO. However, the influence of the presence of vitamin E on the release rate of partially soluble drug, theophylline, was dependent on the MW of PEO; i.e., fast and unstable drug release was obtained in the case of low MW PEO 750 whereas stable drug release was obtained in the case of high MW PEO 303. The release of low water-soluble drug zonisamide was stable regardless of both the presence of vitamin E and the MW of PEO. The presence of vitamin E slightly slowed the release of zonisamide from aged PEO 303 matrices but not PEO 750 matrices. Therefore, in order to achieve a suitable controlled release profile from PEO matrices, not only the presence of vitamin E but also the solubility of the drug and the MW of polyox should be considered.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
α-Tocopherol, ≥95.5%