MilliporeSigma
  • The Epigenetic Reader BRD2 as a Specific Modulator of PAI-1 Expression in Lipopolysaccharide-Stimulated Mouse Primary Astrocytes.
The Epigenetic Reader BRD2 as a Specific Modulator of PAI-1 Expression in Lipopolysaccharide-Stimulated Mouse Primary Astrocytes.
Neurochemical research (2015-09-10)
Chang Soon Choi, Seong Hwi Hong, Seobo Sim, Kyu Suk Cho, Ji-Woon Kim, Sung Min Yang, Se Jin Jeon, Jueng Soo You, Chan Young Shin
ABSTRACT
The post translational modification of lysine acetylation is a key mechanism that regulates chromatin structure. Epigenetic readers, such as the BET domains, are responsible for reading histone lysine acetylation which is a hallmark of open chromatin structure, further providing a scaffold that can be accessed by RNA polymerases as well as transcription factors. Recently, several reports have assessed and highlighted the roles of epigenetic readers in various cellular contexts. However, little is known about their role in the regulation of inflammatory genes, which is critical in exquisitely tuning inflammatory responses to a variety of immune stimuli. In this study, we investigated the role of epigenetic readers BRD2 and BRD4 in the lipopolysaccharide (LPS)-induced immune responses in mouse primary astrocytes. Inflammatory stimulation by LPS showed that the levels of Brd2 mRNA and protein were increased, while Brd4 mRNA levels did not change. Knocking down of Brd2 mRNA using specific small interfering RNA (siRNA) in cultured mouse primary astrocytes inhibited LPS-induced mRNA expression and secretion of plasminogen activator inhibitor-1 (PAI-1). However, no other pro-inflammatory cytokines, such as Il-6, Il-1β and Tnf-α, were affected. Indeed, treatment with bromodomain-containing protein inhibitor, JQ1, blocked Pai-1 mRNA expression through the inhibition of direct BRD2 protein-binding and active histone modification on Pai-1 promoter. Taken together, our data suggest that BRD2 is involved in the modulation of neuroinflammatory responses through PAI-1 and via the regulation of epigenetic reader BET protein, further providing a potential novel therapeutic strategy in neuroinflammatory diseases.
Product Description
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Brd2