MilliporeSigma
Prenatal diagnosis of cystic fibrosis: 10-years experience.
Pathologie-biologie (2015-05-24)
S Hadj Fredj, F Ouali, H Siala, A Bibi, R Othmani, B Dakhlaoui, F Zouari, T Messaoud
ABSTRACT
We present in this study our 10years experience in prenatal diagnosis of cystic fibrosis performed in the Tunisian population. Based on family history, 40 Tunisian couples were selected for prenatal diagnosis. Fetal DNA was isolated from amniotic fluid collected by transabdominal amniocentesis or from chronic villi by transcervical chorionic villus sampling. The genetic analysis for cystic fibrosis mutations was performed by denaturant gradient gel electrophoresis and denaturing high-pressure liquid phase chromatography. We performed microsatellites analysis by capillary electrophoresis in order to verify the absence of maternal cell contamination. Thirteen fetuses were affected, 21 were heterozygous carriers and 15 were healthy with two normal alleles of CFTR gene. Ten couples opted for therapeutic abortion. The microsatellites genotyping showed the absence of contamination of the fetal DNA by maternal DNA in 93.75%. Our diagnostic strategy provides rapid and reliable prenatal diagnosis at risk families of cystic fibrosis.
Sigma-Aldrich
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)