MilliporeSigma
  • MicroRNA-199a-5p Regulates the Proliferation of Pulmonary Microvascular Endothelial Cells in Hepatopulmonary Syndrome.
MicroRNA-199a-5p Regulates the Proliferation of Pulmonary Microvascular Endothelial Cells in Hepatopulmonary Syndrome.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-10-03)
Jing Zeng, Lin Chen, Bing Chen, Kaizhi Lu, Karine Belguise, Xiaobo Wang, Bin Yi
ABSTRACT
Pulmonary microvascular endothelial cell (PMVEC) proliferation and angiogenesis contribute to the development of hepatopulmonary syndrome (HPS). MicroRNA-199a-5p (miR-199a-5p) has emerged as a potent regulator of angiogenesis, and its expression levels significantly decrease in the serum of patients with hepatopathy. However, it has not been reported about whether miR-199a-5p might control PMVEC proliferation. Here, we described the miR-199a-5p governing PMVEC proliferation in HPS. PMVECs were treated with rat serum from common bile duct ligation (CBDL) or sham. MiR-199a-5p mimic or inhibitor was used to change the miR-199a-5p expression. Knockdown of caveolin-1 (Cav-1) was performed using siRNA. NSC-23766 was used to inhibit Rac1 activity. Gene and protein expressions were quantified by qRT-PCR and western blot. Cell proliferation was analyzed by 3H-TdR incorporation and CCK-8 assays. Stress fibers were detected by immunofluorescence. CBDL rat serum induced the down-regulation of miR-199a-5p. Delivery of miR-199a-5p suppressed the CBDL rat serum-induced PMVEC proliferation whereas knockdown of miR-199a-5p promoted PMVEC proliferation. This was accompanied by a decrease and an increase in Cav-1 expression, respectively. Cav-1 siRNA abolished the enhancement of PMVEC proliferation induced by the miR-199a-5p inhibition. Although stress fibers were disrupted in Cav-1 deficient cells, NSC-23766 increased stress fibers and contributed to cell proliferation. CBDL rat serum induced down-regulation of miR-199a-5p in PMVECs, which led to an increase of Cav-1 gene expression. Increased Cav-1 expression, by inhibiting Rac1 activity, led to the formation of stress fibers, which contribute to PMVEC proliferation and thus the pathogenesis of HPS.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cav1
Sigma-Aldrich
MISSION® esiRNA, targeting human CAV1