MilliporeSigma
  • Evaluation of Photoprotective Potential and Percutaneous Penetration by Photoacoustic Spectroscopy of the Schinus terebinthifolius Raddi Extract.
Evaluation of Photoprotective Potential and Percutaneous Penetration by Photoacoustic Spectroscopy of the Schinus terebinthifolius Raddi Extract.
Photochemistry and photobiology (2015-01-13)
Milena K Bulla, Luzmarina Hernandes, Mauro L Baesso, Ana C Nogueira, Antonio C Bento, Bruno B Bortoluzzi, Lara Z Serra, Diogenes A G Cortez
ABSTRACT
Schinus terebinthifolius is a plant rich in phenolic compounds, which have antioxidant properties and can provide new opportunities for treatment and prevention of diseases mediated by ultraviolet radiation like photoaging and skin cancer. The aim of this study was to evaluate the photoprotective potential and ex vivo percutaneous penetration of the crude extract of Schinus terebinthifolius leaves. The extract was tested for antioxidant activity using the 2,2-diphenyl-1-picrylhydrazyl (DPPH) method and β-carotene bleaching test. The sun protection factor was also evaluated. The ex vivo skin permeation of the emulsion and gel formulations were assayed. Fractionation of the extract resulted in gallic acid, ethyl gallate and a mixture of flavonoids, suggesting derivatives of quercetin and myricetin. The phenolic content of the extract was 384.64 ± 2.60 mg GAE g(-1) extract. The antioxidant activity was superior to butylated hydroxytoluene, in DPPH method, and ascorbic acid and rutin, in β-carotene bleaching assay. The extract showed UV absorption with photoprotector potential in the UVB region. The photoacoustic spectroscopy measurements confirmed absorption in the UV region and topical application of the formulations caused no histological changes in the rats' skin. These results suggest that the crude extract of Schinus terebinthifolius leaves may be a promising natural sunscreen product.
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, 97%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG