MilliporeSigma
  • Compact test apparatus for evaluation of flow erosion of marine coatings.
Compact test apparatus for evaluation of flow erosion of marine coatings.
The Review of scientific instruments (2015-11-02)
M A Dębowski, R Quintana, H P Lee
ABSTRACT
An apparatus designed and manufactured for evaluation of flow erosion of coatings or layers is presented in this paper. The setup was primarily designed for coatings intended to perform in dynamic marine environments but can be also used for evaluation using fresh water. The concept is based on an in-line flow test cell and modular design allowing good flexibility of varying testing parameters. The flow rate that can be achieved depends on the flow cell geometry and can reach 28 km/h (15 kn) with the presented setup. Temperature may be adjusted between 15 and 35 °C. Particle and metal ion filters are parts of this setup. The dimensions of the apparatus including all components do not exceed 2 m × 2 m × 2 m. The use of the apparatus is illustrated with the results of evaluation of self-polishing anti-fouling coatings and model, silicon wafer grafted layers.
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
[2-(Methacryloyloxy)ethyl]dimethyl-(3-sulfopropyl)ammonium hydroxide, 95%
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%