MilliporeSigma
  • Conditional promoters for analysis of essential genes in Zymoseptoria tritici.
Conditional promoters for analysis of essential genes in Zymoseptoria tritici.
Fungal genetics and biology : FG & B (2015-06-21)
S Kilaru, W Ma, M Schuster, M Courbot, G Steinberg
ABSTRACT
Development of new fungicides, needed for sustainable control of fungal plant pathogens, requires identification of novel anti-fungal targets. Essential fungal-specific proteins are good candidates, but due to their importance, gene deletion mutants are not viable. Consequently, their cellular role often remains elusive. This hindrance can be overcome by the use of conditional mutants, where expression is controlled by an inducible/repressible promoter. Here, we introduce 5 inducible/repressible promoter systems to study essential genes in the wheat pathogen Zymoseptoria tritici. We fused the gene for enhanced green-fluorescent protein (egfp) to the promoter region of Z. tritici nitrate reductase (Pnar1; induced by nitrogen and repressed by ammonium), 1,4-β-endoxylanase A (Pex1A; induced by xylose and repressed by maltodextrin), l-arabinofuranosidase B (PlaraB; induced by arabinose and repressed by glucose), galactose-1-phosphate uridylyltransferase 7 (Pgal7; induced by galactose and repressed by glucose) and isocitrate lyase (Picl1; induced by sodium acetate and repressed by glucose). This was followed by quantitative analysis of cytoplasmic reporter fluorescence under induced and repressed conditions. We show that Pnar1, PlaraB and Pex1A drive very little or no egfp expression when repressed, but induce moderate protein production when induced. In contrast, Pgal7 and Picl1 show considerable egfp expression when repressed, and were strongly induced in the presence of their inducers. Normalising the expression levels of all promoters to that of the α-tubulin promoter Ptub2 revealed that PlaraB was the weakest promoter (∼20% of Ptub2), whereas Picl1 strongly expressed the reporter (∼250% of Ptub2). The use of these tools promises a better understanding of essential genes, which will help developing novel control strategies that protect wheat from Z. tritici.
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%