MilliporeSigma
  • Comparative Biodistribution and Pharmacokinetic Analysis of Cyclosporine-A in the Brain upon Intranasal or Intravenous Administration in an Oil-in-Water Nanoemulsion Formulation.
Comparative Biodistribution and Pharmacokinetic Analysis of Cyclosporine-A in the Brain upon Intranasal or Intravenous Administration in an Oil-in-Water Nanoemulsion Formulation.
Molecular pharmaceutics (2015-03-19)
Sunita Yadav, Florence Gattacceca, Riccardo Panicucci, Mansoor M Amiji
ABSTRACT
The main objective of this study was to evaluate comparative biodistribution and pharmacokinetics of cyclosporine-A (CsA) following intranasal (IN) administration versus intravenous (IV) administration in Sprague-Dawley rats using an oil-in-water nanoemulsion delivery system. CsA, a hydrophobic peptide that is also a substrate for P-glycoprotein, is a well-known immunosuppressive agent. In the brain, CsA has been shown to be a potent anti-inflammatory and neuroprotective agent. CsA nanoemulsions (CsA-NE) and solution formulations (CsA-S) were prepared using an ultrasonication method and were characterized for drug content, encapsulation efficiency, globule size, and zeta potential. We compared the uptake of CsA-NE and CsA-S in brain regions and peripheral organs following IN and IV administration using LC-MS/MS based bioanalytical method. CsA-NE IN resulted in the highest accumulation compared to that with any other treatment and route of administration; this was consistent for all three regions of brain that were evaluated (olfactory bulbs, mid brain, and hind brain). The brain/blood exposure ratios of 4.49, 0.01, 0.33, and 0.03 for CsA-NE (IN), CsA-NE (IV), CsA-S (IN), and CsA-S (IV), respectively, indicated that CsA-NE is capable of direct nose-to-brain transport, bypassing the blood-brain barrier. Furthermore, CsA-NE administration reduces nontarget organ exposure. These studies show that IN delivery of CsA-NE is an effective way of brain targeting compared to that of other treatment strategies. This approach not only enhances the brain concentration of the peptide but also significantly limits peripheral exposure and the potential for off-target toxicity.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent