MilliporeSigma
The role of cytoplasmic p57 in invasion of hepatocellular carcinoma.
BMC gastroenterology (2015-08-15)
Hui Guo, Yi Li, Tao Tian, Lili Han, Zhiping Ruan, Xuan Liang, Wenjuan Wang, Kejun Nan
ABSTRACT
Our previous research suggested that p57 downregulation could accelerate the growth and invasion of hepatocellular carcinoma in vitro and in vivo. To evaluate the role of cytoplasmic p57 and its regulatory mechanism during hepatocellular carcinoma invasion. We examined the subcellular localization of p57 by immunohistochemistry in 45 pairs of cancerous tissues and adjacent non-cancerous tissues. Moreover, we generated stable p57 knockdown hepatoma cell lines to investigate the mechanism of cytoplasmic p57-mediated regulation of invasion by immunoprecipitation, confocal immunofluorescence microscopy and western blot of nuclear and cytoplasmic extracts. Our results showed that cytoplasmic expression of p57 was reduced in specimens from patients with capsular invasion and metastasis (P < 0.05). Moreover, the level of p-cofilin was decreased in the group lacking cytoplasmic p57 expression (P < 0.05). Co-expression of p57 and p-cofilin was reduced in specimens from patients with tumors at later stages (III + IV), tumors showing capsular invasion and metastatic tumors. We further observed that p57 downregulation decreased the assembly of p57 and LIM domain kinase 1 and its kinase activity, subsequently reducing the level of p-cofilin in the cytoplasm. Cytoplasmic p57 might be a key regulator in hepatocellular carcinoma invasion via the LIM domain kinase 1/p-cofilin pathway.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Coro1a
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdkn1c
Sigma-Aldrich
MISSION® esiRNA, targeting human CORO1A
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%