MilliporeSigma
  • Ferrovibrio xuzhouensis sp. nov., a cyhalothrin-degrading bacterium isolated from cyhalothrin contaminated wastewater.
Ferrovibrio xuzhouensis sp. nov., a cyhalothrin-degrading bacterium isolated from cyhalothrin contaminated wastewater.
Antonie van Leeuwenhoek (2015-05-24)
Man Song, Long Zhang, Bin Sun, Hao Zhang, Hui Ding, Qiang Li, Suhui Guo, Xing Huang
ABSTRACT
A novel cyhalothrin-degrading strain, designated as LM-6(T), was isolated from a cyhalothrin contaminated wastewater sample. The bacterium was found to be Gram stain-negative, non-spore-forming, vibrio-shaped, and motile with a single polar flagellum. Strain LM-6(T) was observed to grow optimally at 28-30 °C, pH 6.0 and in the absence of NaCl. Phylogenetic analysis based on 16S rRNA gene sequence comparisons revealed that strain LM-6(T) is a member of the genus Ferrovibrio, and showed the highest sequence similarity with Ferrovibrio denitrificans Sp-1(T) (97.7 %), followed by Taonella mepensis H1(T) (93.3 %). The major fatty acids of strain LM-6(T) (>5 %) were determined to be C18:1 ω7c and/or C18:1 ω6c, C16:0, C18:1 2-OH and C17:1 iso I and/or anteiso B. The major polar lipids were identified to be phosphatidylglycerol, phosphatidylethanolamine and phosphatidylmethylethanolamine. The major respiratory quinone was determined to be ubiquinone-10. The genomic DNA G+C content of strain LM-6(T) is 66.5 mol %. Strain LM-6(T) showed low DNA-DNA relatedness with F. denitrificans Sp-1(T) (53.1 ± 0.5 %). On the basis of phylogenetic, genomic, phenotypic and chemotaxonomic data, strain LM-6(T) is considered to represent a novel species of the genus Ferrovibrio, for which the name Ferrovibrio xuzhouensis sp. nov. is proposed. The type strain is Ferrovibrio xuzhouensis LM-6(T) (=KCTC 42182(T) = ACCC 19710(T)).
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications