MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • New signaling pathways govern the host response to C. albicans infection in various niches.
New signaling pathways govern the host response to C. albicans infection in various niches.
Genome research (2015-04-11)
Yaoping Liu, Amol C Shetty, Jennifer A Schwartz, L Latey Bradford, Wenjie Xu, Qyunh T Phan, Priti Kumari, Anup Mahurkar, Aaron P Mitchell, Jacques Ravel, Claire M Fraser, Scott G Filler, Vincent M Bruno
ABSTRACT
Candida albicans, the major invasive fungal pathogen of humans, can cause both debilitating mucosal infections and fatal invasive infections. Understanding the complex nature of the host-pathogen interaction in each of these contexts is essential to developing desperately needed therapies to treat fungal infections. RNA-seq enables a systems-level understanding of infection by facilitating comprehensive analysis of transcriptomes from multiple species (e.g., host and pathogen) simultaneously. We used RNA-seq to characterize the transcriptomes of both C. albicans and human endothelial cells or oral epithelial cells during in vitro infection. Network analysis of the differentially expressed genes identified the activation of several signaling pathways that have not previously been associated with the host response to fungal pathogens. Using an siRNA knockdown approach, we demonstrate that two of these pathways-platelet-derived growth factor BB (PDGF BB) and neural precursor-cell-expressed developmentally down-regulated protein 9 (NEDD9)-govern the host-pathogen interaction by regulating the uptake of C. albicans by host cells. Using RNA-seq analysis of a mouse model of hematogenously disseminated candidiasis (HDC) and episodes of vulvovaginal candidiasis (VVC) in humans, we found evidence that many of the same signaling pathways are activated during mucosal (VVC) and/or disseminated (HDC) infections in vivo. Our analyses have uncovered several signaling pathways at the interface between C. albicans and host cells in various contexts of infection, and suggest that PDGF BB and NEDD9 play important roles in this interaction. In addition, these data provide a valuable community resource for better understanding host-fungal pathogen interactions.
Sigma-Aldrich
MISSION® esiRNA, targeting human PDGFRB
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
MISSION® esiRNA, targeting human NEDD9