MilliporeSigma
  • Study on Enhancement Principle and Stabilization for the Luminol-H2O2-HRP Chemiluminescence System.
Study on Enhancement Principle and Stabilization for the Luminol-H2O2-HRP Chemiluminescence System.
PloS one (2015-07-15)
Lihua Yang, Maojun Jin, Pengfei Du, Ge Chen, Chan Zhang, Jian Wang, Fen Jin, Hua Shao, Yongxin She, Shanshan Wang, Lufei Zheng, Jing Wang
ABSTRACT
A luminol-H2O2-HRP chemiluminescence system with high relative luminescent intensity (RLU) and long stabilization time was investigated. First, the comparative study on the enhancement effect of ten compounds as enhancers to the luminol-H2O2-HRP chemiluminescence system was carried out, and the results showed that 4-(imidazol-1-yl)phenol (4-IMP), 4-iodophenol (4-IOP), 4-bromophenol (4-BOP) and 4-hydroxy-4'-iodobiphenyl (HIOP) had the best performance. Based on the experiment, the four enhancers were dissolved in acetone, acetonitrile, methanol, and dimethylformamide (DMF) with various concentrations, the results indicated that 4-IMP, 4-IOP, 4-BOP and HIOP dissolved in DMF with the concentrations of 0.2%, 3.2%, 1.6% and 3.2% could get the highest RLU values. Subsequently, the influences of pH, ionic strength, HRP, 4-IMP, 4-IOP, 4-BOP, HIOP, H2O2 and luminol on the stabilization of the luminol-H2O2-HRP chemiluminescence system were studied, and we found that pH value, ionic strength, 4-IMP, 4-IOP, 4-BOP, HIOP, H2O2 and luminol have little influence on luminescent stabilization, while HRP has a great influence. In different ranges of HRP concentration, different enhancers should be selected. When the concentration is within the range of 0~6 ng/mL, 4-IMP should be selected. When the concentration of HRP ranges from 6 to 25 ng/mL, 4-IOP was the best choice. And when the concentration is within the range of 25~80 ng/mL, HIOP should be selected as the enhancer. Finally, the three well-performing chemiluminescent enhanced solutions (CESs) have been further optimized according to the three enhancers (4-IMP, 4-IOP and HIOP) in their utilized HRP concentration ranges.
Sigma-Aldrich
4-Phenylphenol, 97%
Sigma-Aldrich
4-Phenylphenol, purified by sublimation, 99%
Sigma-Aldrich
4-Methoxyphenol, ReagentPlus®, 99%
Sigma-Aldrich
1-Bromo-4-iodobenzene, 98%
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
2-Iodophenol, 98%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
3-Iodophenol, 98%