MilliporeSigma
Negative regulation of DSS-induced experimental colitis by PILRα.
International immunology (2015-02-25)
Kazuki Kishida, Masako Kohyama, Yosuke Kurashima, Yuta Kogure, Jing Wang, Kouyuki Hirayasu, Tadahiro Suenaga, Hiroshi Kiyono, Jun Kunisawa, Hisashi Arase
ABSTRACT
Inflammatory bowel disease is thought to be a complex multifactorial disease, in which an increased inflammatory response plays an important role. Paired immunoglobulin-like type 2 receptor α (PILRα), well conserved in almost all mammals, is an inhibitory receptor containing immunoreceptor tyrosine-based inhibitory motifs in the cytoplasmic domain. PILRα is mainly expressed on myeloid cells and plays an important role in the regulation of inflammation. In the present study, we investigated the function of PILRα in inflammatory bowel disease using PILRα-deficient mice. When mice were orally administered dextran sulfate sodium (DSS), colonic mucosal injury and inflammation were significantly exacerbated in DSS-treated PILRα-deficient mice compared with wild-type (WT) mice. Flow cytometric analysis revealed that neutrophil and macrophage cell numbers were higher in the colons of DSS-treated PILRα-deficient mice than in those of WT mice. Blockade of CXCR2 expressed on neutrophils using a CXCR2 inhibitor decreased the severity of colitis observed in PILRα-deficient mice. These results suggest that PILRα negatively regulates inflammatory colitis by regulating the infiltration of inflammatory cells such as neutrophils and macrophages.
Sigma-Aldrich
o-Dianisidine, peroxidase substrate