MilliporeSigma
  • Determination of residual arsenic compounds in chicken muscle by ultra-performance liquid chromatography coupled with ultraviolet detection after pre-column derivatization with toluene-3,4-dithiol.
Determination of residual arsenic compounds in chicken muscle by ultra-performance liquid chromatography coupled with ultraviolet detection after pre-column derivatization with toluene-3,4-dithiol.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-11-10)
Han Young Eom, Dong-Hyug Yang, Joon Hyuk Suh, Unyong Kim, Junghyun Kim, Hyun-Deok Cho, Sang Beom Han
ABSTRACT
A simple and sensitive derivatization method using toluene-3,4-dithiol as a derivatization reagent for the simultaneous analysis of seven arsenic compounds (roxarsone, nitarsone, p-arsanilic acid, o-arsanilic acid, phenylarsonic acid, phenylarsine oxide, and mono-methylarsonic acid) in chicken muscle was developed and validated by ultra-performance liquid chromatography coupled with ultraviolet detection (UPLC-UV). The structure of the derivatized arsenic compounds was confirmed by liquid chromatography-ion trap mass spectrometry or gas chromatography-mass spectrometry. Optimization of the derivatization reaction conditions was carried out by investigating the influence of reagent concentration, buffer or additive acids, temperature, and time. The optimized conditions were a derivatization reagent concentration of 20mg/mL with 0.05mol/L HCl as an additive acid at 60°C for 15min. In this study, baseline separation of arsenic compounds could be achieved within 13min, except for phenylarsonic acid and phenylarsine oxide whose derivatized products are equal. The developed method was successfully validated and applied to 12 chicken muscle samples from Korean districts and other countries.
Sigma-Aldrich
p-Arsanilic acid
Sigma-Aldrich
Ammonium acetate solution, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Arsenic, powder, ≥99.997% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Arsenic, 99.999% trace metals basis
Sigma-Aldrich
Toluene-3,4-dithiol, technical grade, 90%
Sigma-Aldrich
Phenylarsine oxide, ≥97%, powder
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%