MilliporeSigma
  • TGF-β Blood Levels Distinguish Between Influenza A (H1N1)pdm09 Virus Sepsis and Sepsis due to Other Forms of Community-Acquired Pneumonia.
TGF-β Blood Levels Distinguish Between Influenza A (H1N1)pdm09 Virus Sepsis and Sepsis due to Other Forms of Community-Acquired Pneumonia.
Viral immunology (2015-04-30)
Erick J Rendón-Ramirez, Alejandro Ortiz-Stern, Corazon Martinez-Mejia, Mario C Salinas-Carmona, Adrian Rendon, Viviana L Mata-Tijerina, Adrian G Rosas-Taraco
ABSTRACT
There is a strong interest in finding adequate biomarkers to aid in the diagnosis and prognosis of influenza A (H1N1)pdm09 virus infection. In this study, serum levels of inflammatory cytokines and laboratory markers were evaluated to assess their usefulness as biomarkers of influenza A (H1N1)pdm09 and their association with fatal cases. Serum samples of consecutive patients with a clinical presentation suggestive of influenza A (H1N1)pdm09 and progression to sepsis were evaluated. Serum inflammatory cytokines and routine laboratory tests were performed and correlated with positivity for influenza A (H1N1)pdm09 influenza by real time reverse transcription polymerase chain reaction and the results of three clinical severity scores (Sequential Organ Failure Assessment [SOFA], CURB-65, and Acute Physiology and Chronic Health Evaluation II [APACHE II]). High SOFA scores and some of its individual components, but not CURB-65 or APACHE II scores, correlate with fatal cases regardless of etiology. Total and unconjugated bilirubin, Ca(++), Cl(-), prothrombin times, and partial thromboplastin times discriminate influenza A (H1N1)pdm09 from other causes of community-acquired pneumonia. High levels of IL-8, IL-10, and IL-17 were increased in influenza A (H1N1)pdm09 patients when compared with controls (p<0.05). IL-6 levels were significantly elevated in influenza A (H1N1)pdm09 patients and non-(H1N1)pdm09 patients when compared with controls (p<0.05). TGF-β serum levels discern between healthy controls, influenza A (H1N1)pdm09 patients, and patients with other causes of community-acquired pneumonia. TGF-β levels were negatively correlated with SOFA on admission in influenza A (H1N1)pdm09 patients. TGF-β levels are a useful tool for differentiating influenza A (H1N1)pdm09 from other causes of pneumonia progressing to sepsis.
Sigma-Aldrich
Iodotrimethylsilane, 97%