MilliporeSigma
  • Proteomic identification of alpha-2-HS-glycoprotein as a plasma biomarker of hypopharyngeal squamous cell carcinoma.
Proteomic identification of alpha-2-HS-glycoprotein as a plasma biomarker of hypopharyngeal squamous cell carcinoma.
International journal of clinical and experimental pathology (2015-10-16)
Wen-Dong Tian, Jun-Zheng Li, Shui-Wang Hu, Xiao-Wei Peng, Gang Li, Xiong Liu, Huai-Hong Chen, Xia Xu, Xiang-Ping Li
ABSTRACT
Hypopharyngeal squamous cell carcinoma (HSCC) has very poor prognosis compared with other head and neck squamous cell carcinomas. Late-stage diagnosis of HSCC increases mortality. Therefore, more effective biomarkers for early diagnosis of HSCC are necessary. Unfortunately, appropriate biomarkers for clinical diagnosis and prognosis have not been identified yet. However, recent progresses in quantitative proteomics have offered opportunities to identify plasma proteins as biomarkers for HSCC. In the present study, plasma samples were analyzed by two-dimensional differential gel electrophoresis (2D-DIGE), and differentially expressed proteins were identified by matrix assisted laser desorption ionization-time of flight/time of flight mass spectrometry (MALDI-TOF/TOF MS). A total of 26 proteins representing 12 unique gene products were identified. The up-regulation proteins were alpha-2-HS-glycoprotein (AHSG), complement C4-B, haptoglobin, C-reactive protein, and ceruloplasmin, whereas the down-regulation proteins were serum albumin, angiotensinogen, alpha-1-antichymotrypsin, Ig gamma-3 chain C region, fibrinogen gamma chain, apolipoprotein A-I, and Ig kappa chain C region. Among all the differentially expressed proteins, AHSG was validated by western blot and ELISA. The results were consistent with the data from 2D-DIGE, further suggesting that AHSG may be employed as a potential biomarker for the early diagnosis of HSCC. In summary, this study was the first to use 2D-DIGE and MALDI-TOF/TOF platform to identify the potential plasma biomarkers for HSCC. The plasma AHSG showed great potential for HSCC screening.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%