MilliporeSigma
  • Inhibition of pro-protein convertase subtilisin/kexin type 6 has a protective role against synovitis in a rat model of rheumatoid arthritis.
Inhibition of pro-protein convertase subtilisin/kexin type 6 has a protective role against synovitis in a rat model of rheumatoid arthritis.
Molecular medicine reports (2015-10-16)
Huiyu Jiang, Lin Wang, Jihong Pan
ABSTRACT
The aim of the present study was to assess the effects of pro-protein convertase subtilisin/kexin type 6 (PCSK6), a proteinase implicated in the proteolytic activity of various precursor proteins and involved in the regulation of protein maturation, in fibroblast‑like synoviocytes (FLS) of a rat model of collagen‑induced arthritis (CIA). Cultured FLS from CIA models were subjected to small interfering RNA mediated PCSK6 knockdown, followed by assessment of the proliferation, invasive and migratory capacity, the secretion of inflammation factors and the cell cycle. Expression of genes associated with proliferation, invasion, migration and inflammation was detected by reverse transcription polymerase chain reaction. The results showed that PCSK6 knockdown significantly decreased the cell proliferation, invasion and migration of FLS from rats with CIA. ELISA showed an obvious decrease of tumor necrosis factor α and interleukin 1β secretion, and flow cytometric analysis revealed G0/G1 arrest of FLS following PCSK6 knockdown. Furthermore, a decrease in the mRNA levels of inflammation‑associated chemokine CXCL9, angiogenesis‑associated genes MMP‑2, MMP‑9 and NOSTRIN, hypoxia‑associated gene HIF‑1α, adhesion‑associated gene MPZL2, proliferation‑associated gene IGF‑2 and citrullination‑associated gene PADI4 was detected after PCSK6 knockdown. The results of the present study indicated that inhibition of PCSK6 may have a protective role against synovitis in rheumatoid arthritis.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pcsk6
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PCSK6 (2)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)