MilliporeSigma
  • Up to 100% Improvement in Dynamic Nuclear Polarization Solid-State NMR Sensitivity Enhancement of Polymers by Removing Oxygen.
Up to 100% Improvement in Dynamic Nuclear Polarization Solid-State NMR Sensitivity Enhancement of Polymers by Removing Oxygen.
Macromolecular rapid communications (2015-05-27)
Dao Le, Fabio Ziarelli, Trang N T Phan, Giulia Mollica, Pierre Thureau, Fabien Aussenac, Olivier Ouari, Didier Gigmes, Paul Tordo, Stéphane Viel
ABSTRACT
High-field dynamic nuclear polarization (DNP) has emerged as a powerful technique for improving the sensitivity of solid-state NMR (SSNMR), yielding significant sensitivity enhancements for a variety of samples, including polymers. Overall, depending upon the type of polymer, the molecular weight, and the DNP sample preparation method, sensitivity enhancements between 5 and 40 have been reported. These promising enhancements remain, however, far from the theoretical maximum (>1000). Crucial to the success of DNP SSNMR is the DNP signal enhancement (εDNP ), which is the ratio of the NMR signal intensities with and without DNP. It is shown here that, for polymers exhibiting high affinity toward molecular oxygen (e.g., polystyrene), removing part of the absorbed (paramagnetic) oxygen from the solid-state samples available as powders (instead of dissolved or dispersed in a solvent) increases proton nuclear relaxation times and εDNP, hereby providing up to a two-fold sensitivity increase (i.e., a four-fold reduction in experimental time).
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized