MilliporeSigma
  • Comparison of platinum chromium everolimus-eluting stent with cobalt chromium everolimus-eluting stent in unselected patients undergoing percutaneous coronary intervention.
Comparison of platinum chromium everolimus-eluting stent with cobalt chromium everolimus-eluting stent in unselected patients undergoing percutaneous coronary intervention.
European review for medical and pharmacological sciences (2015-07-15)
R-G Li, C-H Lee, A Low, M Chan, K H Chan, A M Richards, X-K Qu, W-Y Fang, H C Tan
ABSTRACT
The recent PLATINUM trial has demonstrated that the use of the new generation platinum chromium everolimus-eluting stents (PtCr-EES) yield clinical outcomes similar to those obtained by the use of cobalt chromium everolimus-eluting stents (CoCr-EES) in selected patients with 1 or 2 de novo coronary artery lesions. This study aimed to compare the safety and efficacy of the PtCr-EES and CoCr-EES in unselected patients from a real-life single-center registry. From July 2009 through November 2010, 788 consecutive patients in our institution with symptomatic coronary artery disease who were treated with the CoCr-EES (n = 410) or PtCr-EES (n = 378) were enrolled into this study. The primary endpoint of the study was target-lesion failure (TLF) at 12-month follow-up and the secondary endpoints were major adverse cardiovascular events and stent thrombosis. The prevalence of TLF in the PtCr-EES group (4.5%) was similar to that in the CoCr-EES group (3.9%). In addition, there were no significant differences in the 12-month rates of cardiac death (2.1% vs. 1.5%), myocardial infarction (2.4% vs. 3.9%), ischemia-driven target lesion revascularization (2.4% vs. 2.2%), and definite or probable stent thrombosis (0.5% vs. 1.5%, all p > 0.05). At 12-month follow-up, the PtCr-EES is comparable in safety and efficacy to the CoCr-EES in unselected patients with coronary artery diseases.
Sigma-Aldrich
Butyl methacrylate, 99%, contains monomethyl ether hydroquinone as inhibitor