MilliporeSigma
  • Biosynthesis of poly(3-hydroxybutyrate-co-3-hydroxyvalerate) by Halogeometricum borinquense strain E3.
Biosynthesis of poly(3-hydroxybutyrate-co-3-hydroxyvalerate) by Halogeometricum borinquense strain E3.
International journal of biological macromolecules (2015-04-22)
Bhakti B Salgaonkar, Judith M Bragança
ABSTRACT
Polyhydroxyalkanoates (PHA's) can be a key solution for pollution problems caused by plastics derived from petrochemical sources. Extremely halophilic archaeon Halogeometricum borinquense strain E3 showed maximum PHA accumulation of 73.51 ±1.7% (wt/wt) of cell dry weight (CDW) with 2% glucose. The crotonic acid assay, XRD, FT-IR and (1)H NMR analysis revealed that the polymer was a co-polymer of poly(3-hydroxybutyrate-co-3-hydroxyvalerate) [P(HB-co-HV)] comprising of 21.47% HV units. This is the first report on P(HB-co-HV) production by an extremely halophilic archaeon Hgm. borinquense strain E3.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer